Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACGTACTCGGGGGGCACCTGCGC[A/G]TCGGGCGCCGCCTTGCGGTAGAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606481 MIM: 610918 MIM: 602127 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5J PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INPP5J - inositol polyphosphate-5-phosphatase J | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SELM - selenoprotein M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080430.2 | 423 | Silent Mutation | GAC,GAT | D,D 121 | NP_536355.1 |
SMTN - smoothelin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207017.1 | 423 | Intron | NP_001193946.1 | |||
NM_001207018.1 | 423 | Intron | NP_001193947.1 | |||
NM_006932.4 | 423 | Intron | NP_008863.3 | |||
NM_134269.2 | 423 | Intron | NP_599031.1 | |||
NM_134270.2 | 423 | Intron | NP_599032.2 | |||
XM_006724298.1 | 423 | Intron | XP_006724361.1 | |||
XM_006724302.1 | 423 | Intron | XP_006724365.1 | |||
XM_011530333.2 | 423 | Intron | XP_011528635.1 | |||
XM_011530336.2 | 423 | Intron | XP_011528638.1 | |||
XM_011530337.2 | 423 | Intron | XP_011528639.1 | |||
XM_011530338.2 | 423 | Intron | XP_011528640.1 | |||
XM_011530339.2 | 423 | Intron | XP_011528641.1 | |||
XM_011530340.2 | 423 | Intron | XP_011528642.1 | |||
XM_011530341.1 | 423 | Intron | XP_011528643.1 | |||
XM_017028913.1 | 423 | Intron | XP_016884402.1 | |||
XM_017028914.1 | 423 | Intron | XP_016884403.1 | |||
XM_017028915.1 | 423 | Intron | XP_016884404.1 | |||
XM_017028916.1 | 423 | Intron | XP_016884405.1 | |||
XM_017028917.1 | 423 | Intron | XP_016884406.1 | |||
XM_017028918.1 | 423 | Intron | XP_016884407.1 | |||
XM_017028919.1 | 423 | Intron | XP_016884408.1 |