Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTCCGGCTCCTGGAGAACATGG[C/T]CCGGCCTCCCGGGGGCTCTGGTCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605648 | ||||||||||||||||||||
Literature Links: |
FBXO7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO7 - F-box protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033024.1 | 130 | Missense Mutation | GCC,GTC | A,V 2 | NP_001028196.1 | |
NM_001257990.1 | 130 | UTR 5 | NP_001244919.1 | |||
NM_012179.3 | 130 | Intron | NP_036311.3 | |||
XM_011530106.1 | 130 | Intron | XP_011528408.1 |