Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTCTGCGATGCAGCTGTTGGGC[A/G]CAGCAGGCCATGGGTTACCACGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 146770 | ||||||||||||||||||||
Literature Links: |
DRICH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DRICH1 - aspartate rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016449.3 | 476 | Intron | NP_057533.2 | |||
XM_011530206.1 | 476 | Intron | XP_011528508.1 | |||
XM_011530207.2 | 476 | Intron | XP_011528509.1 | |||
XM_011530208.2 | 476 | Intron | XP_011528510.1 | |||
XM_011530209.2 | 476 | Intron | XP_011528511.1 | |||
XM_011530210.2 | 476 | Intron | XP_011528512.1 | |||
XM_017028813.1 | 476 | Intron | XP_016884302.1 |
IGLL1 - immunoglobulin lambda like polypeptide 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020070.3 | 476 | Missense Mutation | CGC,TGC | R,C 40 | NP_064455.1 | |
NM_152855.2 | 476 | Missense Mutation | CGC,TGC | R,C 40 | NP_690594.1 | |
XM_011530169.2 | 476 | Missense Mutation | CGC,TGC | R,C 40 | XP_011528471.1 |