Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACAGCCCGGGCCTCAGCTGGCCC[C/G]GTGTTCAAGGGCGTCTGTAAGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601786 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSDC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSDC2 - cold shock domain containing C2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014460.3 | 498 | Silent Mutation | CCC,CCG | P,P 67 | NP_055275.1 |
PMM1 - phosphomannomutase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |