Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCCATGAAAGGCATGTTGACGG[C/T]ACCACGGATATGGCCCGAGTCCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602496 MIM: 180370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPST PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MPST - mercaptopyruvate sulfurtransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX33 - testis expressed 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TST - thiosulfate sulfurtransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270483.1 | 665 | Missense Mutation | ACC,GCC | T,A 208 | NP_001257412.1 | |
NM_003312.5 | 665 | Missense Mutation | ACC,GCC | T,A 208 | NP_003303.2 |