Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAGAAACCCGATGGAGGCAATG[A/T]ATCCACACATATTCTACTTCCACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608993 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOBEC3F PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOBEC3F - apolipoprotein B mRNA editing enzyme catalytic subunit 3F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006666.1 | 879 | Intron | NP_001006667.1 | |||
NM_145298.5 | 879 | Missense Mutation | AAT,TAT | N,Y 196 | NP_660341.2 | |
XM_011529994.2 | 879 | Missense Mutation | AAT,TAT | N,Y 265 | XP_011528296.1 | |
XM_017028642.1 | 879 | Intron | XP_016884131.1 | |||
XM_017028643.1 | 879 | Intron | XP_016884132.1 |