Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTACCAGTGCTCCTCGGCGGGGCT[A/G]AACTGCTGGTGCCACCGGAAACAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GAB4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GAB4 - GRB2 associated binding protein family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037814.1 | 921 | Missense Mutation | NP_001032903.1 | |||
XM_011546114.2 | 921 | Missense Mutation | XP_011544416.1 | |||
XM_011546115.1 | 921 | Missense Mutation | XP_011544417.1 | |||
XM_011546116.2 | 921 | Intron | XP_011544418.1 | |||
XM_011546117.1 | 921 | Missense Mutation | XP_011544419.1 | |||
XM_017028575.1 | 921 | Missense Mutation | XP_016884064.1 | |||
XM_017028576.1 | 921 | Missense Mutation | XP_016884065.1 |