Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACTGTGGCGAACATCAGGGCAC[C/T]GGCAGCGTTAGACTTCACATGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 139160 MIM: 605663 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNAZ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNAZ - G protein subunit alpha z | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RSPH14 - radial spoke head 14 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014433.2 | 1117 | Missense Mutation | AGT,GGT | S,G 253 | NP_055248.1 | |
XM_011530149.2 | 1117 | Missense Mutation | AGT,GGT | S,G 253 | XP_011528451.1 | |
XM_011530150.1 | 1117 | Missense Mutation | AGT,GGT | S,G 253 | XP_011528452.1 | |
XM_011530151.2 | 1117 | Missense Mutation | AGT,GGT | S,G 253 | XP_011528453.1 | |
XM_011530152.2 | 1117 | Missense Mutation | AGT,GGT | S,G 253 | XP_011528454.1 | |
XM_011530154.2 | 1117 | Missense Mutation | AGT,GGT | S,G 185 | XP_011528456.1 | |
XM_011530155.2 | 1117 | Missense Mutation | AGT,GGT | S,G 121 | XP_011528457.1 | |
XM_017028774.1 | 1117 | Missense Mutation | AGT,GGT | S,G 253 | XP_016884263.1 |