Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGACGTGAATTTCTGATGAAAG[C/T]TATCTTACCAAGTTTAAATTCATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601754 | ||||||||||||||||||||
Literature Links: |
C22orf39 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C22orf39 - chromosome 22 open reading frame 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UFD1L - ubiquitin fusion degradation 1 like (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035247.2 | 1061 | Intron | NP_001030324.2 | |||
NM_005659.6 | 1061 | Intron | NP_005650.2 | |||
XM_011530361.2 | 1061 | Missense Mutation | ACT,GCT | T,A 264 | XP_011528663.1 |