Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCTGGAGGAATGCTTGGAGAAGC[A/C]CCTGCCGCTCCCCGACTTGCAGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606673 | ||||||||||||||||||||
Literature Links: |
ADORA2A-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADORA2A-AS1 - ADORA2A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UPB1 - beta-ureidopropionase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016327.2 | 92 | Missense Mutation | CAC,CCC | H,P 16 | NP_057411.1 | |
XM_011530222.2 | 92 | Missense Mutation | CAC,CCC | H,P 16 | XP_011528524.1 | |
XM_011530223.2 | 92 | Missense Mutation | CAC,CCC | H,P 16 | XP_011528525.1 | |
XM_011530224.2 | 92 | Missense Mutation | CAC,CCC | H,P 16 | XP_011528526.1 | |
XM_011530225.2 | 92 | UTR 5 | XP_011528527.1 | |||
XM_017028825.1 | 92 | Missense Mutation | CAC,CCC | H,P 16 | XP_016884314.1 | |
XM_017028826.1 | 92 | Missense Mutation | CAC,CCC | H,P 16 | XP_016884315.1 | |
XM_017028827.1 | 92 | Missense Mutation | CAC,CCC | H,P 16 | XP_016884316.1 | |
XM_017028828.1 | 92 | Missense Mutation | CAC,CCC | H,P 16 | XP_016884317.1 |