Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGACCCAGGATGGGGCCAGATA[C/T]TCCCTGTAAATGACTCAAGTCCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603952 MIM: 607445 | ||||||||||||||||||||
Literature Links: |
DRG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DRG1 - developmentally regulated GTP binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EIF4ENIF1 - eukaryotic translation initiation factor 4E nuclear import factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164501.1 | 3095 | Missense Mutation | ATA,GTA | I,V 865 | NP_001157973.1 | |
NM_001164502.1 | 3095 | Missense Mutation | ATA,GTA | I,V 691 | NP_001157974.1 | |
NM_019843.3 | 3095 | Missense Mutation | ATA,GTA | I,V 865 | NP_062817.2 | |
XM_005261686.2 | 3095 | Missense Mutation | ATA,GTA | I,V 866 | XP_005261743.1 | |
XM_005261687.2 | 3095 | Missense Mutation | ATA,GTA | I,V 866 | XP_005261744.1 | |
XM_005261688.2 | 3095 | Missense Mutation | ATA,GTA | I,V 866 | XP_005261745.1 | |
XM_006724281.3 | 3095 | Missense Mutation | ATA,GTA | I,V 907 | XP_006724344.3 | |
XM_011530280.1 | 3095 | Missense Mutation | ATA,GTA | I,V 865 | XP_011528582.1 | |
XM_011530281.2 | 3095 | Missense Mutation | ATA,GTA | I,V 866 | XP_011528583.1 | |
XM_017028863.1 | 3095 | Missense Mutation | ATA,GTA | I,V 906 | XP_016884352.1 | |
XM_017028864.1 | 3095 | Missense Mutation | ATA,GTA | I,V 865 | XP_016884353.1 |