Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCGGGCCTGGCAGAGTCTGGCCA[A/G]CCACGCCGTGGCAGAGGAACCCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608666 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEX26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PEX26 - peroxisomal biogenesis factor 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127649.2 | 409 | Missense Mutation | AAC,AGC | N,S 67 | NP_001121121.1 | |
NM_001199319.1 | 409 | Missense Mutation | AAC,AGC | N,S 67 | NP_001186248.1 | |
NM_017929.5 | 409 | Missense Mutation | AAC,AGC | N,S 67 | NP_060399.1 |