Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCGGGATGCTTGCTCTGCAGAA[C/T]CTGGATGATGCATTCCGAGATGAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 160000 | ||||||||||||||||||||
Literature Links: |
MB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MB - myoglobin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005368.2 | 515 | Missense Mutation | ATT,GTT | I,V 115 | NP_005359.1 | |
NM_203377.1 | 515 | Missense Mutation | ATT,GTT | I,V 115 | NP_976311.1 | |
NM_203378.1 | 515 | Missense Mutation | ATT,GTT | I,V 115 | NP_976312.1 | |
XM_005261605.2 | 515 | Missense Mutation | ATT,GTT | I,V 115 | XP_005261662.1 |