Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTTCTGCATCCAAGATAGCCTT[A/G]GCACCATGCAGCCTGGGCGGGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 617034 MIM: 610020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GATSL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GATSL3 - GATS protein like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D10A - TBC1 domain family member 10A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204240.1 | 1276 | Silent Mutation | GCC,GCT | A,A 397 | NP_001191169.1 | |
NM_031937.2 | 1276 | Silent Mutation | GCC,GCT | A,A 390 | NP_114143.1 |