Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCGGCCCCCAGGGGGGCAGAGG[C/T]GCCGCCACCATGAGCCTGGGCAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606084 MIM: 150571 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC42EP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC42EP1 - CDC42 effector protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152243.2 | 445 | Silent Mutation | GGC,GGT | G,G 9 | NP_689449.1 | |
XM_006724107.1 | 445 | Silent Mutation | GGC,GGT | G,G 9 | XP_006724170.1 | |
XM_006724108.1 | 445 | Silent Mutation | GGC,GGT | G,G 9 | XP_006724171.1 | |
XM_006724109.1 | 445 | Silent Mutation | GGC,GGT | G,G 9 | XP_006724172.1 | |
XM_017028541.1 | 445 | Silent Mutation | GGC,GGT | G,G 9 | XP_016884030.1 |
LGALS2 - galectin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |