Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGGTAGAGCTCGCGCACCCTCCG[C/T]TTGGCCTCGTTCATGTCCCGACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602138 MIM: 615588 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDUFA6 - NADH:ubiquinone oxidoreductase subunit A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002490.3 | 227 | Silent Mutation | AAA,AAG | K,K 55 | NP_002481.2 |
NDUFA6-AS1 - NDUFA6 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMDT1 - single-pass membrane protein with aspartate rich tail 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |