Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTGCACTACCACCCGGACTCGCC[G/T]GCCAAGGGCGCGCAGTGGATGAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602054 | ||||||||||||||||||||
Literature Links: |
TBX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBX1 - T-box 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005992.1 | 864 | Silent Mutation | CCG,CCT | P,P 200 | NP_005983.1 | |
NM_080646.1 | 864 | Silent Mutation | CCG,CCT | P,P 200 | NP_542377.1 | |
NM_080647.1 | 864 | Silent Mutation | CCG,CCT | P,P 200 | NP_542378.1 | |
XM_006724312.2 | 864 | Silent Mutation | CCG,CCT | P,P 200 | XP_006724375.1 | |
XM_017028925.1 | 864 | Silent Mutation | CCG,CCT | P,P 250 | XP_016884414.1 | |
XM_017028926.1 | 864 | Silent Mutation | CCG,CCT | P,P 200 | XP_016884415.1 | |
XM_017028927.1 | 864 | Intron | XP_016884416.1 | |||
XM_017028928.1 | 864 | Silent Mutation | CCG,CCT | P,P 250 | XP_016884417.1 |