Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAGCCTCGCCACCTTGGGGGCC[A/G]GGCCATCATCACCAAGAGCTTTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 100850 | ||||||||||||||||||||
Literature Links: |
ACO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACO2 - aconitase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098.2 | 2073 | Missense Mutation | CAG,CGG | Q,R 684 | NP_001089.1 | |
XM_017028812.1 | 2073 | Missense Mutation | CAG,CGG | Q,R 651 | XP_016884301.1 |
POLR3H - RNA polymerase III subunit H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018050.3 | 2073 | UTR 3 | NP_001018060.1 | |||
NM_001018052.3 | 2073 | UTR 3 | NP_001018062.1 | |||
NM_001282884.1 | 2073 | UTR 3 | NP_001269813.1 | |||
NM_001282885.1 | 2073 | UTR 3 | NP_001269814.1 | |||
NM_138338.4 | 2073 | UTR 3 | NP_612211.1 | |||
XM_011529993.1 | 2073 | Intron | XP_011528295.1 | |||
XM_017028641.1 | 2073 | Intron | XP_016884130.1 |