Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCCCTATCCCTACAGGATGTGT[G/T]CCCCTGTGATGGTGGAGCTGGAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604414 MIM: 602229 | ||||||||||||||||||||
Literature Links: |
MIR6820 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6820 - microRNA 6820 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR2F - RNA polymerase II subunit F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301129.1 | 354 | Missense Mutation | GCC,TCC | A,S 68 | NP_001288058.1 | |
NM_001301130.1 | 354 | Missense Mutation | GCC,TCC | A,S 77 | NP_001288059.1 | |
NM_001301131.1 | 354 | Missense Mutation | GCC,TCC | A,S 77 | NP_001288060.1 | |
NM_021974.4 | 354 | Missense Mutation | GCC,TCC | A,S 77 | NP_068809.1 | |
XM_017028832.1 | 354 | Missense Mutation | GCC,TCC | A,S 77 | XP_016884321.1 |
SOX10 - SRY-box 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |