Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCATAGCATTTTTCTGGTGCCAC[C/T]GCCCTACAGCGAGCAGGAGAAGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TEX33 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TEX33 - testis expressed 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163857.1 | 876 | Missense Mutation | CAG,CGG | Q,R 250 | NP_001157329.1 | |
NM_178552.3 | 876 | Missense Mutation | CAG,CGG | Q,R 165 | NP_848647.1 | |
XM_011530165.2 | 876 | Missense Mutation | CAG,CGG | Q,R 250 | XP_011528467.1 | |
XM_011530166.1 | 876 | Missense Mutation | CAG,CGG | Q,R 165 | XP_011528468.1 |