Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCTTTCTTCCCTTTTTACGCAA[A/T]GACTGTCCTTCTCCAGAGAAAGCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601754 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C22orf39 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C22orf39 - chromosome 22 open reading frame 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UFD1L - ubiquitin fusion degradation 1 like (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035247.2 | 1153 | Intron | NP_001030324.2 | |||
NM_005659.6 | 1153 | Intron | NP_005650.2 | |||
XM_011530361.2 | 1153 | Silent Mutation | TCA,TCT | S,S 294 | XP_011528663.1 |