Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTTGGCTCAGTGCCTAAAGTGTC[C/T]GCAATAAGTAAAACGCAAACAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614140 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPECC1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPECC1L - sperm antigen with calponin homology and coiled-coil domains 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145468.3 | 353 | Silent Mutation | TCC,TCT | S,S 15 | NP_001138940.3 | |
NM_001254732.2 | 353 | Silent Mutation | TCC,TCT | S,S 15 | NP_001241661.2 | |
NM_001254733.1 | 353 | Intron | NP_001241662.1 | |||
NM_015330.4 | 353 | Silent Mutation | TCC,TCT | S,S 15 | NP_056145.4 |
SPECC1L-ADORA2A - SPECC1L-ADORA2A readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |