Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCATGGAGGTCATGGGGGTCAC[A/G]GGGGTCATGGGATTCACGGGCCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610409 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAIAP2L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAIAP2L2 - BAI1 associated protein 2 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025045.5 | 1611 | Silent Mutation | CCC,CCT | P,P 395 | NP_079321.3 | |
XM_005261751.4 | 1611 | Silent Mutation | CCC,CCT | P,P 395 | XP_005261808.1 | |
XM_011530379.2 | 1611 | Silent Mutation | CCC,CCT | P,P 395 | XP_011528681.1 | |
XM_011530380.2 | 1611 | Silent Mutation | CCC,CCT | P,P 377 | XP_011528682.1 | |
XM_011530381.2 | 1611 | Silent Mutation | CCC,CCT | P,P 376 | XP_011528683.1 | |
XM_011530382.2 | 1611 | Silent Mutation | CCC,CCT | P,P 376 | XP_011528684.1 | |
XM_011530383.2 | 1611 | Silent Mutation | CCC,CCT | P,P 371 | XP_011528685.1 | |
XM_011530384.2 | 1611 | Silent Mutation | CCC,CCT | P,P 365 | XP_011528686.1 | |
XM_011530386.2 | 1611 | Silent Mutation | CCC,CCT | P,P 200 | XP_011528688.1 | |
XM_011530387.2 | 1611 | Silent Mutation | CCC,CCT | P,P 200 | XP_011528689.1 | |
XM_011530388.2 | 1611 | Silent Mutation | CCC,CCT | P,P 188 | XP_011528690.1 |
SLC16A8 - solute carrier family 16 member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |