Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAAGGTGGTCACTTCTCATAAA[A/G]CGTTTTGAACATTCTGGACAAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601804 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SP3 - Sp3 transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017371.4 | 2589 | Silent Mutation | CGC,CGT | R,R 621 | NP_001017371.3 | |
NM_001172712.1 | 2589 | Silent Mutation | CGC,CGT | R,R 686 | NP_001166183.1 | |
NM_003111.4 | 2589 | Silent Mutation | CGC,CGT | R,R 689 | NP_003102.1 |