Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGGCTAAGTATCCATTTTTGGA[A/G]TGGATCTGATGGGTTGCATGACACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601756 | ||||||||||||||||||||
Literature Links: |
GALNT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GALNT3 - polypeptide N-acetylgalactosaminyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004482.3 | 2140 | Missense Mutation | CTC,TTC | L,F 624 | NP_004473.2 | |
XM_005246449.1 | 2140 | Missense Mutation | CTC,TTC | L,F 624 | XP_005246506.1 | |
XM_011510929.1 | 2140 | Missense Mutation | CTC,TTC | L,F 624 | XP_011509231.1 | |
XM_017003770.1 | 2140 | Missense Mutation | CTC,TTC | L,F 624 | XP_016859259.1 |