Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGTAGACCAAAGGATTCACTCC[C/T]GAGGAAACATAGCCTATCCACACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601122 | ||||||||||||||||||||
Literature Links: |
HTR2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HTR2B - 5-hydroxytryptamine receptor 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000867.4 | 1574 | Silent Mutation | TCA,TCG | S,S 373 | NP_000858.3 | |
NM_001320758.1 | 1574 | Silent Mutation | TCA,TCG | S,S 331 | NP_001307687.1 | |
XM_005246520.3 | 1574 | Silent Mutation | TCA,TCG | S,S 254 | XP_005246577.1 | |
XM_006712482.2 | 1574 | Silent Mutation | TCA,TCG | S,S 348 | XP_006712545.1 |
PSMD1 - proteasome 26S subunit, non-ATPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191037.1 | 1574 | Intron | NP_001177966.1 | |||
NM_002807.3 | 1574 | Intron | NP_002798.2 | |||
XM_017004517.1 | 1574 | Intron | XP_016860006.1 |