Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCTCATTGCAAAATCTTTGTAG[C/T]TGCTGGTACCATCCTCCAGCTCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611208 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPLL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HNRNPLL - heterogeneous nuclear ribonucleoprotein L like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142650.1 | 1612 | Missense Mutation | AAC,AGC | N,S 421 | NP_001136122.1 | |
NM_138394.3 | 1612 | Missense Mutation | AAC,AGC | N,S 426 | NP_612403.2 | |
XM_005264639.3 | 1612 | Missense Mutation | AAC,AGC | N,S 426 | XP_005264696.2 | |
XM_005264640.3 | 1612 | Missense Mutation | AAC,AGC | N,S 421 | XP_005264697.2 | |
XM_011533165.2 | 1612 | Missense Mutation | AAC,AGC | N,S 402 | XP_011531467.1 | |
XM_011533166.2 | 1612 | Missense Mutation | AAC,AGC | N,S 426 | XP_011531468.1 |