Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAAAGCAGCTCTAATGACGAAGA[A/C]GAGAACAACCGAAGAAAGATCTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612636 | ||||||||||||||||||||
Literature Links: |
UNC80 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UNC80 - unc-80 homolog, NALCN activator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032504.1 | 590 | Silent Mutation | GAA,GAC | E,D 170 | NP_115893.1 | |
NM_182587.3 | 590 | Silent Mutation | GAA,GAC | E,D 170 | NP_872393.3 | |
XM_005246476.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_005246533.1 | |
XM_011511004.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_011509306.1 | |
XM_011511005.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_011509307.1 | |
XM_011511006.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_011509308.1 | |
XM_011511007.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_011509309.1 | |
XM_011511008.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_011509310.1 | |
XM_011511010.2 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_011509312.1 | |
XM_011511012.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_011509314.1 | |
XM_017003884.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_016859373.1 | |
XM_017003885.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_016859374.1 | |
XM_017003886.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_016859375.1 | |
XM_017003887.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_016859376.1 | |
XM_017003888.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_016859377.1 | |
XM_017003889.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_016859378.1 | |
XM_017003890.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_016859379.1 | |
XM_017003891.1 | 590 | Missense Mutation | GAA,GAC | E,D 52 | XP_016859380.1 | |
XM_017003892.1 | 590 | Intron | XP_016859381.1 | |||
XM_017003893.1 | 590 | Missense Mutation | GAA,GAC | E,D 170 | XP_016859382.1 | |
XM_017003894.1 | 590 | Intron | XP_016859383.1 |