Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTATAGCCTCCATGACTTTAAA[C/T]GGAGGGGACTGGAATCTGCTTGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606704 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR75 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR75 - G protein-coupled receptor 75 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006794.3 | 1933 | Missense Mutation | ATT,GTT | I,V 540 | NP_006785.1 |
GPR75-ASB3 - GPR75-ASB3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164165.1 | 1933 | Intron | NP_001157637.1 |
MIR3682 - microRNA 3682 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |