Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAACTGAGCAAACCCAATCAGCTT[C/T]ACTTCATCAGGAATCTCCGACCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607648 | ||||||||||||||||||||
Literature Links: |
STK39 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STK39 - serine/threonine kinase 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013233.2 | 1762 | Silent Mutation | GTA,GTG | V,V 534 | NP_037365.2 | |
XM_005246465.2 | 1762 | Silent Mutation | GTA,GTG | V,V 513 | XP_005246522.1 | |
XM_017003813.1 | 1762 | Intron | XP_016859302.1 | |||
XM_017003814.1 | 1762 | Intron | XP_016859303.1 | |||
XM_017003815.1 | 1762 | Silent Mutation | GTA,GTG | V,V 432 | XP_016859304.1 | |
XM_017003816.1 | 1762 | Intron | XP_016859305.1 | |||
XM_017003817.1 | 1762 | Silent Mutation | GTA,GTG | V,V 370 | XP_016859306.1 |