Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACACAGACATGCAGCAGTTGGCC[C/T]GGGAGACCTCCGTGGACCCAGACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 182125 | ||||||||||||||||||||
Literature Links: |
SPR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPR - sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003124.4 | 678 | Missense Mutation | CGG,TGG | R,W 210 | NP_003115.1 |