Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGGTTCAACTGCTCATCATAGC[C/G]ATAGTAGTGGGCTTTAGAGTGGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607164 MIM: 610231 MIM: 604240 | ||||||||||||||||||||
Literature Links: |
LBX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LBX2 - ladybird homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LBX2-AS1 - LBX2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCGF1 - polycomb group ring finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032673.2 | 516 | Missense Mutation | CGC,GGC | R,G 164 | NP_116062.2 |
TLX2 - T-cell leukemia homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |