Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTGCACATGGTTAAGATGGGAC[A/T]TCATTTGACAAGGGCTGGTGTTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602507 | ||||||||||||||||||||
Literature Links: |
LOC105374845 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105374845 - uncharacterized LOC105374845 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF103 - ring finger protein 103 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198951.1 | 2049 | Missense Mutation | GAA,GAT | E,D 678 | NP_001185880.1 | |
NM_001198952.1 | 2049 | Intron | NP_001185881.1 | |||
NM_005667.3 | 2049 | Missense Mutation | GAA,GAT | E,D 682 | NP_005658.1 |
RNF103-CHMP3 - RNF103-CHMP3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198954.1 | 2049 | Intron | NP_001185883.1 |