Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTGTGGCCGAGTACATGAAGGA[C/G]GTGTTGACTAATCCAGCCAAGGTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612828 MIM: 615898 | ||||||||||||||||||||
Literature Links: |
CEBPZ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEBPZ - CCAAT/enhancer binding protein zeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005760.2 | 270 | Intron | NP_005751.2 |
NDUFAF7 - NADH:ubiquinone oxidoreductase complex assembly factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083946.1 | 270 | Missense Mutation | GAC,GAG | D,E 65 | NP_001077415.1 | |
NM_144736.4 | 270 | Missense Mutation | GAC,GAG | D,E 65 | NP_653337.1 | |
XM_005264410.1 | 270 | Missense Mutation | GAC,GAG | D,E 23 | XP_005264467.1 | |
XM_005264411.1 | 270 | Missense Mutation | GAC,GAG | D,E 65 | XP_005264468.1 | |
XM_011532956.1 | 270 | UTR 5 | XP_011531258.1 |