Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCATTAGCAAGCTCCTGGACTT[G/T]TTTCTGGGCCAGGAGATTCGCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607805 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNNM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNNM4 - cyclin and CBS domain divalent metal cation transport mediator 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020184.3 | 993 | Missense Mutation | TTG,TTT | L,F 326 | NP_064569.3 | |
XM_005263914.3 | 993 | Missense Mutation | TTG,TTT | L,F 326 | XP_005263971.1 | |
XM_005263915.3 | 993 | Missense Mutation | TTG,TTT | L,F 326 | XP_005263972.1 | |
XM_011510955.2 | 993 | Missense Mutation | TTG,TTT | L,F 326 | XP_011509257.1 | |
XM_011510956.2 | 993 | Missense Mutation | TTG,TTT | L,F 326 | XP_011509258.1 | |
XM_017003799.1 | 993 | Intron | XP_016859288.1 |