Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACTCACTTTACTACATATACAGC[A/C]CATACACCACCATGCTTAAGATGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614029 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPDYA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPDYA - speedy/RINGO cell cycle regulator family member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT61B - tRNA methyltransferase 61B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017910.3 | 806 | Missense Mutation | TGG,TGT | W,C 356 | NP_060380.3 | |
XM_005264393.2 | 806 | Missense Mutation | TGG,TGT | W,C 356 | XP_005264450.1 | |
XM_006712037.2 | 806 | Missense Mutation | TGG,TGT | W,C 258 | XP_006712100.1 | |
XM_017004401.1 | 806 | Missense Mutation | TGG,TGT | W,C 258 | XP_016859890.1 | |
XM_017004402.1 | 806 | Intron | XP_016859891.1 | |||
XM_017004403.1 | 806 | Missense Mutation | TGG,TGT | W,C 120 | XP_016859892.1 |