Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCTGGGGGCTCACAAGTGGGAGC[A/G]GTAATTGCGATTGGCTGTGGGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190231 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928278 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101928278 - uncharacterized LOC101928278 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105373876 - uncharacterized LOC105373876 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNP1 - transition protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003284.3 | 179 | Missense Mutation | CGC,TGC | R,C 52 | NP_003275.1 |