Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCCATTTATAGATGTTTACATG[C/G]TCCTTTCTATCAAGGGACTTCATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALS2CR11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALS2CR11 - amyotrophic lateral sclerosis 2 chromosome region candidate 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168216.1 | 4645 | Intron | NP_001161688.1 | |||
NM_001168217.1 | 4645 | Intron | NP_001161689.1 | |||
NM_001168221.1 | 4645 | Missense Mutation | GAC,GAG | D,E 1531 | NP_001161693.1 | |
NM_152525.5 | 4645 | Intron | NP_689738.3 | |||
XM_006712331.3 | 4645 | Intron | XP_006712394.1 | |||
XM_006712332.3 | 4645 | Intron | XP_006712395.1 | |||
XM_006712333.3 | 4645 | Intron | XP_006712396.1 | |||
XM_006712334.3 | 4645 | Intron | XP_006712397.1 | |||
XM_006712335.3 | 4645 | Intron | XP_006712398.1 | |||
XM_006712336.3 | 4645 | Intron | XP_006712399.1 | |||
XM_011510736.2 | 4645 | Intron | XP_011509038.1 |