Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGCACGTACCGTATTCTAGTAT[C/T]TAACAATTCCTTAATCATTGCCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608100 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFU1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NFU1 - NFU1 iron-sulfur cluster scaffold | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002755.2 | 828 | Missense Mutation | AAT,GAT | N,D 178 | NP_001002755.1 | |
NM_001002756.2 | 828 | Missense Mutation | AAT,GAT | N,D 37 | NP_001002756.1 | |
NM_015700.3 | 828 | Missense Mutation | AAT,GAT | N,D 154 | NP_056515.2 | |
XM_017003808.1 | 828 | Intron | XP_016859297.1 |