Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCATGTTTTCTGCTTGAGAGGT[A/G]CTGATGTACCAGTTGGGGAACTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147720 | ||||||||||||||||||||
Literature Links: |
IL1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL1B - interleukin 1 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000576.2 | 804 | Silent Mutation | AGC,AGT | S,S 239 | NP_000567.1 | |
XM_017003988.1 | 804 | Silent Mutation | AGC,AGT | S,S 208 | XP_016859477.1 |