Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTCACATCATACATTGCAGGCC[A/G]GGATTTCTTTCCAAACCTGAAATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607299 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNER PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNER - delta/notch like EGF repeat containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139072.3 | 2130 | Missense Mutation | CGG,TGG | R,W 707 | NP_620711.3 | |
XM_005246950.2 | 2130 | Missense Mutation | CGG,TGG | R,W 666 | XP_005247007.1 |