Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAACCCCCCATTGTTGAAAGTCA[C/T]GTGGTCTTTGAGACAATTTACAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601828 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NR4A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NR4A2 - nuclear receptor subfamily 4 group A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006186.3 | 3746 | Missense Mutation | ATG,GTG | M,V 539 | NP_006177.1 | |
XM_005246621.3 | 3746 | Missense Mutation | ATG,GTG | M,V 550 | XP_005246678.1 | |
XM_005246622.3 | 3746 | Missense Mutation | ATG,GTG | M,V 476 | XP_005246679.1 | |
XM_006712553.3 | 3746 | Missense Mutation | ATG,GTG | M,V 525 | XP_006712616.1 | |
XM_011511246.1 | 3746 | Missense Mutation | CAT,CGT | H,R 515 | XP_011509548.1 | |
XM_017004219.1 | 3746 | Missense Mutation | ATG,GTG | M,V 539 | XP_016859708.1 | |
XM_017004220.1 | 3746 | Missense Mutation | ATG,GTG | M,V 514 | XP_016859709.1 |