Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGTAAGGAATGGAGATTCTTGTA[C/T]AATCTGTAAAATTCAAAAAATCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604891 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCKAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCKAP1 - NCK associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013436.4 | 3521 | Missense Mutation | ATA,GTA | I,V 1092 | NP_038464.1 | |
NM_205842.2 | 3521 | Missense Mutation | ATA,GTA | I,V 1098 | NP_995314.1 | |
XM_006712200.3 | 3521 | Missense Mutation | ATA,GTA | I,V 1096 | XP_006712263.1 | |
XM_006712201.3 | 3521 | Missense Mutation | ATA,GTA | I,V 1090 | XP_006712264.1 |