Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAGATCTTAGTAATTACAGTTA[C/T]AGCTCTACCCTGCCCCCTTTTCTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 146928 | ||||||||||||||||||||
Literature Links: |
CXCR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CXCR2 - C-X-C motif chemokine receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168298.1 | 156 | Silent Mutation | TAC,TAT | Y,Y 25 | NP_001161770.1 | |
NM_001557.3 | 156 | Silent Mutation | TAC,TAT | Y,Y 25 | NP_001548.1 | |
XM_005246530.3 | 156 | Intron | XP_005246587.1 | |||
XM_017003990.1 | 156 | Silent Mutation | TAC,TAT | Y,Y 25 | XP_016859479.1 | |
XM_017003991.1 | 156 | Silent Mutation | TAC,TAT | Y,Y 25 | XP_016859480.1 | |
XM_017003992.1 | 156 | Silent Mutation | TAC,TAT | Y,Y 25 | XP_016859481.1 |