Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCTCCCACCTTCTGTTGTGGCTA[A/C]TGTTGGTGGTAAAATTTTCACATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616865 | ||||||||||||||||||||
Literature Links: |
PAPOLG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAPOLG - poly(A) polymerase gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022894.3 | 495 | Missense Mutation | AAT,ACT | N,T 91 | NP_075045.2 | |
XM_005264500.3 | 495 | Missense Mutation | AAT,ACT | N,T 91 | XP_005264557.1 | |
XM_005264501.1 | 495 | Missense Mutation | AAT,ACT | N,T 47 | XP_005264558.1 | |
XM_011533041.1 | 495 | UTR 5 | XP_011531343.1 | |||
XM_017004723.1 | 495 | UTR 5 | XP_016860212.1 | |||
XM_017004724.1 | 495 | UTR 5 | XP_016860213.1 |