Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATATGTCCATCTGTACCAACCTC[C/T]CCAGCCGCTGGACCGTGTTCCAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602886 | ||||||||||||||||||||
Literature Links: |
GPR39 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR39 - G protein-coupled receptor 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001508.2 | 1109 | Missense Mutation | CCC,TCC | P,S 214 | NP_001499.1 | |
XM_011511021.2 | 1109 | Missense Mutation | CCC,TCC | P,S 214 | XP_011509323.1 |