Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAAGATATGAGAAACATGGAGGT[G/T]TTCTGACCCTGCTTCCAGAAAATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 177010 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERPINE2 - serpin family E member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136528.1 | 1055 | Missense Mutation | AAC,CAC | N,H 332 | NP_001130000.1 | |
NM_001136530.1 | 1055 | Missense Mutation | AAC,CAC | N,H 344 | NP_001130002.1 | |
NM_006216.3 | 1055 | Missense Mutation | AAC,CAC | N,H 333 | NP_006207.1 | |
XM_005246641.2 | 1055 | Missense Mutation | AAC,CAC | N,H 345 | XP_005246698.1 | |
XM_017004329.1 | 1055 | Missense Mutation | AAC,CAC | N,H 333 | XP_016859818.1 | |
XM_017004330.1 | 1055 | Missense Mutation | AAC,CAC | N,H 333 | XP_016859819.1 | |
XM_017004331.1 | 1055 | Missense Mutation | AAC,CAC | N,H 332 | XP_016859820.1 | |
XM_017004332.1 | 1055 | Missense Mutation | AAC,CAC | N,H 332 | XP_016859821.1 |