Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAAATCCACTCCAACATCGACC[A/G]GGGCTTTGGAGATCTGAGCTGACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 102720 MIM: 605556 | ||||||||||||||||||||
Literature Links: |
DPP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPP4 - dipeptidyl peptidase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001935.3 | 2729 | Missense Mutation | CCG,CTG | P,L 723 | NP_001926.2 | |
XM_005246371.2 | 2729 | Missense Mutation | CCG,CTG | P,L 722 | XP_005246428.1 |
SLC4A10 - solute carrier family 4 member 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |