Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCACCCAGTGCTCATGGTCACC[A/G]GCTTCGTCTTCATCCAGGGCATCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605745 | ||||||||||||||||||||
Literature Links: |
CYBRD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYBRD1 - cytochrome b reductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127383.1 | 359 | Missense Mutation | AGC,GGC | S,G 57 | NP_001120855.1 | |
NM_001256909.1 | 359 | Intron | NP_001243838.1 | |||
NM_024843.3 | 359 | Missense Mutation | AGC,GGC | S,G 57 | NP_079119.3 |