Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCTTGCAGCCCGGTTTGGGGAT[C/G]TGGTCCTTGCTGCTCTGCGGGTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147810 MIM: 604512 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1R1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL1R1 - interleukin 1 receptor type 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL1RL2 - interleukin 1 receptor like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003854.2 | 264 | Missense Mutation | ATC,ATG | I,M 0 | NP_003845.2 | |
XM_005264035.1 | 264 | Missense Mutation | ATC,ATG | I,M 29 | XP_005264092.1 | |
XM_005264036.1 | 264 | Missense Mutation | ATC,ATG | I,M 1 | XP_005264093.1 | |
XM_006712820.2 | 264 | Missense Mutation | TCT,TGT | S,C 79 | XP_006712883.1 | |
XM_006712822.2 | 264 | Intron | XP_006712885.1 | |||
XM_011512091.1 | 264 | Missense Mutation | ATC,ATG | I,M 29 | XP_011510393.1 | |
XM_011512092.1 | 264 | Missense Mutation | ATC,ATG | I,M 29 | XP_011510394.1 | |
XM_011512093.1 | 264 | Missense Mutation | ATC,ATG | I,M 29 | XP_011510395.1 | |
XM_011512094.1 | 264 | Missense Mutation | ATC,ATG | I,M 1 | XP_011510396.1 | |
XM_011512096.1 | 264 | Missense Mutation | ATC,ATG | I,M 29 | XP_011510398.1 | |
XM_017005174.1 | 264 | Missense Mutation | ATC,ATG | I,M 29 | XP_016860663.1 | |
XM_017005175.1 | 264 | Missense Mutation | TCT,TGT | S,C 79 | XP_016860664.1 | |
XM_017005176.1 | 264 | Missense Mutation | TCT,TGT | S,C 79 | XP_016860665.1 | |
XM_017005177.1 | 264 | UTR 5 | XP_016860666.1 | |||
XM_017005178.1 | 264 | UTR 5 | XP_016860667.1 | |||
XM_017005179.1 | 264 | UTR 5 | XP_016860668.1 | |||
XM_017005180.1 | 264 | UTR 5 | XP_016860669.1 |